Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039520.2 | NP_001034609.1 | 5 - 6 | 880 | 75 | |
NM_001253893.1 | NP_001240822.1 | 5 - 6 | 880 | 75 | ||
NM_001253894.1 | NP_001240823.1 | 5 - 6 | 880 | 75 | ||
NM_007871.2 | NP_031897.2 | 5 - 6 | 880 | 75 | ||
GenBank mRNA | AK005012.2 | - | 5 - 6 | 856 | 75 | |
AK150046.1 | - | 5 - 6 | 843 | 75 | ||
AK154375.1 | - | 4 - 5 | 544 | 75 | ||
AK157844.1 | - | 5 - 6 | 850 | 75 | ||
AK170574.1 | - | 5 - 6 | 856 | 75 | ||
AK171049.1 | - | 5 - 6 | 834 | 75 | ||
AK171566.1 | - | 5 - 6 | 844 | 75 | ||
AK172231.1 | - | 5 - 6 | 861 | 75 | ||
AK180287.1 | - | 2 - 3 | 197 | 75 | ||
AK206038.1 | - | 2 - 3 | 131 | 75 | ||
L31398.1 | - | 5 - 6 | 820 | 75 |
Set Membership: |
![]() ![]() ![]() ![]() |