Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039520.2 | NP_001034609.1 | 8 - 9 | 1319 | 93 | |
NM_001253893.1 | NP_001240822.1 | 8 - 9 | 1319 | 93 | ||
NM_001253894.1 | NP_001240823.1 | 8 - 9 | 1319 | 93 | ||
NM_007871.2 | NP_031897.2 | 8 - 9 | 1319 | 93 | ||
GenBank mRNA | AK005012.2 | - | 8 - 9 | 1295 | 93 | |
AK015410.1 | - | 7 - 8 | 984 | 93 | ||
AK150046.1 | - | 8 - 9 | 1282 | 93 | ||
AK154375.1 | - | 7 - 8 | 983 | 93 | ||
AK157844.1 | - | 8 - 9 | 1289 | 93 | ||
AK170574.1 | - | 8 - 9 | 1295 | 93 | ||
AK171049.1 | - | 8 - 9 | 1273 | 93 | ||
AK171566.1 | - | 8 - 9 | 1283 | 93 | ||
AK172231.1 | - | 8 - 9 | 1300 | 93 | ||
AK198303.1 | - | 1 - 2 | 51 | 93 | ||
AK208713.1 | - | 2 - 3 | 156 | 93 | ||
AK213385.1 | - | 2 - 3 | 180 | 93 | ||
L31398.1 | - | 8 - 9 | 1259 | 93 |
Set Membership: |
![]() ![]() ![]() ![]() |