Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01151967_g1
          See other ZFP653 GE Assays ›
          Gene Symbol:
          Zfp653
          Gene Name:
          zinc finger protein 653
          Gene Aliases:
          E430039K05Rik, Znf653
          Chromosome Location:
          Chr.9: 22055460 - 22071370 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01151967_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp653
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177318.2 NP_796292.1 8 - 9 1689 106
          GenBank mRNA AK046492.1 - 6 - 7 2057 106
          AK089104.1 - 8 - 9 1501 106
          AK157635.1 - 6 - 7 2057 106
          AY072705.1 - 8 - 9 1699 106
          BC038037.1 - 8 - 9 1689 4190337 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001895.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp653

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350250 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350324 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350334 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350332 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035023 BC038037.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035023 AK046492.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035023 NM_177318.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104950 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109650 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173182 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169717 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035023 BC038037.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035023 AK046492.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035023 NM_177318.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398141 AY072705.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398141 BC038037.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398141 AK046492.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398141 AK089104.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398141 AK157635.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26350249 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169717 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173182 - -
          Whole Mouse Genome G4122A A_51_P398141 AY072705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398141 BC038037.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398141 AK046492.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398141 AK089104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398141 AK157635.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138925 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134687 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169717 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109650 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08073242 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08111535 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350324 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350250 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350330 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350334 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26350332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26350193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07464544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07823669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07708241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08036481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08193179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08065930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08193177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07784339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07823665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07823684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07703507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07823664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07464546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07893211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08168102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08057859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26350257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07461923 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591718 - -
          Affymetrix HT MG-430 PM Array Plate 1435481_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10591715 - -
          Mouse Genome 430 2.0 Array 1435481_at - -
          Mouse Genome 430 2.0 Array 1435481_PM_at - -
          Mouse Genome 430 2.0 Array 1455560_at - -
          Mouse Genome 430 2.0 Array 1455560_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319601_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319601_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900015982.mm.1 TC0900001895.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900015982.mm.1 NM_177318.3 Refseq
          Mouse Expression Set 430 1435481_PM_at - -
          Mouse Expression Set 430 1435481_at - -
          Murine Genome U74v2 Set 104666_PM_at - -
          Murine Genome U74v2 Set 104666_at - -
          Affymetrix HT Mouse Genome 430B Array 1435481_at - -
          Affymetrix HT Mouse Genome 430B Array 1435481_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 724265 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610717 - -
          Illumina mouse-6 v1.0 expression beadchip 610717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755621 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755621 - -
          mouse-6 v1.1 (Illumina) scl0319601.1_34-S - -
          Illumina Sentrix mouse-6.v1 bead array 610717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0319601.1_34-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2755621 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755621 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0319601.1_34-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0