Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01157852_m1
          See other NDUFB2 GE Assays ›
          Gene Symbol:
          Ndufb2
          Gene Name:
          NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2
          Gene Aliases:
          1810011O01Rik, 8kDa, AGGG, AI325567, CI-AGGG
          Chromosome Location:
          Chr.6: 39592551 - 39599472 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01157852_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ndufb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026612.3 NP_080888.1 3 - 4 371 101

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ndufb2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24383894 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24383895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24383892 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24383891 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119420 AK019008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119420 AK028334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119420 BC013510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119420 NM_026612.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119420 AK007495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P13910 AK028334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P13910 BC013510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P13910 AK007495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P13910 AK010642.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P13910 NM_026612.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41835 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171116 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120452 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119420 AK019008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119420 AK028334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119420 BC013510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119420 NM_026612.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119420 AK007495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P13910 AK028334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P13910 BC013510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P13910 AK007495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P13910 AK010642.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P13910 NM_026612.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361184 AK019008.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24383884 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171116 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05871757 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120452 - -
          Whole Mouse Genome G4122A A_51_P361184 AK019008.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115042 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118670 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120452 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171116 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05376109 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05376113 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05376120 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05376114 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05376117 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05376106 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24383891 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24383895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24383894 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24383892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24383938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05091237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05376106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05376109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05376116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05015299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05376101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05865738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05141288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05272850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05141285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05272849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05376100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05259433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05141293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05141287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05141296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24383897 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537456 - -
          Affymetrix HT MG-430 PM Array Plate 1416834_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1453806_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448483_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1442658_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10537452 - -
          Mouse Genome 430 2.0 Array 1416834_PM_x_at - -
          Mouse Genome 430 2.0 Array 1442658_at - -
          Mouse Genome 430 2.0 Array 1442658_PM_at - -
          Mouse Genome 430 2.0 Array 1453806_PM_at - -
          Mouse Genome 430 2.0 Array 1448483_a_at - -
          Mouse Genome 430 2.0 Array 1453806_at - -
          Mouse Genome 430 2.0 Array 1416834_x_at - -
          Mouse Genome 430 2.0 Array 1448483_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68198_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68198_at - -
          Mouse Expression Set 430 1416834_PM_x_at - -
          Mouse Expression Set 430 1416834_x_at - -
          Mouse Expression Set 430 1448483_a_at - -
          Mouse Expression Set 430 1453806_PM_at - -
          Mouse Expression Set 430 1448483_PM_a_at - -
          Mouse Expression Set 430 1453806_at - -
          Mouse Expression Set 430 1442658_PM_at - -
          Mouse Expression Set 430 1442658_at - -
          Murine Genome U74v2 Set 95131_f_at - -
          Murine Genome U74v2 Set 162473_PM_r_at - -
          Murine Genome U74v2 Set 95132_r_at - -
          Murine Genome U74v2 Set 95132_PM_r_at - -
          Murine Genome U74v2 Set 160342_PM_r_at - -
          Murine Genome U74v2 Set 160342_r_at - -
          Murine Genome U74v2 Set 95131_PM_f_at - -
          Murine Genome U74v2 Set 162473_r_at - -
          Murine Genome U74v2 Set 169599_at - -
          Murine Genome U74v2 Set 171505_f_at - -
          Murine Genome U74v2 Set 169599_PM_at - -
          Murine Genome U74v2 Set 171505_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1453806_PM_at - -
          Mouse Genome 430A 2.0 Array 1416834_x_at - -
          Mouse Genome 430A 2.0 Array 1448483_a_at - -
          Mouse Genome 430A 2.0 Array 1453806_at - -
          Mouse Genome 430A 2.0 Array 1448483_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416834_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448483_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453806_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1453806_at - -
          Affymetrix HT Mouse Genome 430A Array 1416834_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1416834_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448483_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1442658_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442658_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103450707 - -
          MouseWG-6 v2 Expression BeadChip 101690601 - -
          Illumina mouse-6 v1.0 expression beadchip 103450707 - -
          Illumina mouse-6 v1.0 expression beadchip 1230750 - -
          Illumina mouse-6 v1.0 expression beadchip 101690601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636435 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2447445 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243847 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636435 - -
          mouse-6 v1.1 (Illumina) scl29105.1.1_14-S - -
          mouse-6 v1.1 (Illumina) scl0068198.2_271-S - -
          mouse-6 v1.1 (Illumina) scl30167.5_55-S - -
          Illumina Sentrix mouse-6.v1 bead array 103450707 - -
          Illumina Sentrix mouse-6.v1 bead array 1230750 - -
          Illumina Sentrix mouse-6.v1 bead array 101690601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0068198.2_271-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101690601 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230750 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103450707 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2636435 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243847 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2636435 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2447445 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0