Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500018990.mm.1 |
TC0500002084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018990.mm.1 |
NR_033188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018990.mm.1 |
NR_033188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018991.mm.1 |
TC0500002084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018991.mm.1 |
NM_177676.6.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018991.mm.1 |
NM_177676.6 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018991.mm.1 |
NR_033188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018991.mm.1 |
NR_033188.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018993.mm.1 |
TC0500002084.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500018993.mm.1 |
NR_033188.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500018993.mm.1 |
NR_033188.1 |
Refseq |