Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01162447_s1
          See other HDGFL1 GE Assays ›
          Gene Symbol:
          Hdgfl1
          Gene Name:
          hepatoma derived growth factor-like 1
          Gene Aliases:
          4921520N01Rik, HRP-1, Hdgfrp1, Pwwp1
          Chromosome Location:
          Chr.13: 26768172 - 26770165 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01162447_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hdgfl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008232.3 NP_032258.2 1 1871 109
          GenBank mRNA AF180109.1 - 1 1719 109
          AK133357.1 - 1 1877 109
          BC108388.1 - 1 1821 30920044 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001683.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hdgfl1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 D63663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 AF180109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 AK133357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 BC108388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 NM_008232.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 D63663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 AF180109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 AK133357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 BC108388.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230298 NM_008232.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338154 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164006 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 D63663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 AF180109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 AK133357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 BC108388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 NM_008232.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 D63663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 AF180109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 AK133357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 BC108388.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230298 NM_008232.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28899233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164006 - -
          Whole Mouse Genome G4122A A_51_P230298 D63663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 AF180109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 AK133357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 BC108388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 NM_008232.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 D63663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 AF180109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 AK133357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 BC108388.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230298 NM_008232.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133094 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164006 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998401 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998410 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998397 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998370 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998399 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10998379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28899224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10841558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10565170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10841559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10720198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11037636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10961607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10946953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10581301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10961602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28899211 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408369 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408368 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408370 - -
          Affymetrix HT MG-430 PM Array Plate 1421294_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1421293_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408367 - -
          Mouse Genome 430 2.0 Array 1421293_at - -
          Mouse Genome 430 2.0 Array 1421293_PM_at - -
          Mouse Genome 430 2.0 Array 1421294_PM_at - -
          Mouse Genome 430 2.0 Array 1421294_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15192_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15192_at - -
          Affymetrix Mouse Clariom S Array PSR1300010467.mm.1 TC1300001683.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300010467.mm.1 NM_008232.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300010467.mm.1 NM_008232.3 Refseq
          Mouse Expression Set 430 1421293_PM_at - -
          Mouse Expression Set 430 1421293_at - -
          Mouse Expression Set 430 1421294_at - -
          Mouse Expression Set 430 1421294_PM_at - -
          Murine Genome U74v2 Set 99869_PM_at - -
          Murine Genome U74v2 Set 99869_at - -
          Murine Genome U74v2 Set 170852_at - -
          Murine Genome U74v2 Set 168214_r_at - -
          Murine Genome U74v2 Set 170852_PM_at - -
          Murine Genome U74v2 Set 168214_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1421293_PM_at - -
          Mouse Genome 430A 2.0 Array 1421294_at - -
          Mouse Genome 430A 2.0 Array 1421293_at - -
          Mouse Genome 430A 2.0 Array 1421294_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421294_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421293_at - -
          Affymetrix HT Mouse Genome 430A Array 1421294_at - -
          Affymetrix HT Mouse Genome 430A Array 1421293_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 509813 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4070707 - -
          Illumina mouse-6 v1.0 expression beadchip 4070707 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004070707 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2751905 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2751905 - -
          mouse-6 v1.1 (Illumina) scl44174.1.1_69-S - -
          Illumina Sentrix mouse-6.v1 bead array 4070707 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44174.1.1_69-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070707 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2874380 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2874380 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl44174.1.1_69-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0