Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164638_m1
          See other 4930527G23RIK GE Assays ›
          Gene Symbol:
          4930527G23Rik
          Gene Name:
          RIKEN cDNA 4930527G23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 29442200 - 29466107 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164638_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930527G23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045395.1 3 - 4 454 85
          GenBank mRNA AK019704.1 - 3 - 4 455 85
          BY715476.1 - 3 - 4 451 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV267612.1 - 1 - 2 145 Chr. 18 - 29452123 - 29466107 [+] on Build GRCm38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930578E11 gene 4930578E11Rik Anti-sense Chr. 18 - 29396716 - 29496416 on NCBI Build GRCm38
          Downstream phosphoinositide-3-kinase,
          class 3
          Pik3c3 5330434F23Rik,
          Vps34
          806.0kb Sense Chr. 18 - 30272094 - 30348126 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000230.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13921392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13679872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13962415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13957839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14019033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13957842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13962420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13957850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13731571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13957851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13921366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13813210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13957826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13777718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31587506 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800001616.mm.1 TC1800000230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800001616.mm.1 NR_045395.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001616.mm.1 NR_045395.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001618.mm.1 TC1800000230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800001618.mm.1 NR_045395.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001618.mm.1 NR_045395.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001615.mm.1 TC1800000230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800001615.mm.1 NR_045395.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001615.mm.1 NR_045395.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001621.mm.1 TC1800000230.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800001621.mm.1 NR_045395.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800001621.mm.1 NR_045395.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 409186 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0