Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164813_m1
          See other 4930528H21RIK GE Assays ›
          Gene Symbol:
          4930528H21Rik
          Gene Name:
          RIKEN cDNA 4930528H21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 4031842 - 4047611 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164813_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930528H21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK019707.1 - 2 - 3 288 85
          BY715486.1 - 2 - 3 288 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream guanine nucleotide binding protein (G protein),
          gamma 11
          Gng11 0610037B21Rik -23.4kb Anti-sense Chr. 6 - 4003987 - 4008445 on NCBI Build GRCm38
          Downstream blocked early in transport 1 homolog (S. cerevisiae) Bet1 AW555236,
          Bet-1
          31.9kb Sense Chr. 6 - 4076904 - 4086995 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109786 AK019707.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126966 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109786 AK019707.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126966 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05847486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05731683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05462017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05731684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05610704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P05754143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24202222 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0