Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164929_g1
          See other FASTK GE Assays ›
          Gene Symbol:
          Fastk
          Gene Name:
          Fas-activated serine/threonine kinase
          Gene Aliases:
          0610011K02Rik, 610011K02Rik
          Chromosome Location:
          Chr.5: 24441039 - 24445282 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164929_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fastk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023229.2 NP_075718.2 2 - 3 580 64
          GenBank mRNA AF168682.1 - 1 - 2 475 64
          AK002552.1 - 2 - 3 531 64
          AK038596.1 - 2 - 3 532 64
          AK052667.1 - 2 - 3 525 64
          AK139374.1 - 1 - 2 1298 64
          AK151859.1 - 2 - 3 532 64
          AK152369.1 - 2 - 3 581 64
          AK159751.1 - 2 - 3 532 64
          AK164317.1 - 539 64
          AK167346.1 - 1 - 2 1292 64
          AK191049.1 - 1 - 2 43 64
          AK195958.1 - 1 - 2 43 64
          BC013547.1 - 2 - 3 497 4218374 64

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fastk

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          apoptotic process
          phosphorylation
          regulation of RNA splicing
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          Fas-activated serine/threonine kinase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500018920.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018920.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018920.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018929.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018929.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018929.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018921.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018921.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018921.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018923.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018923.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018923.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018916.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018916.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018916.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018917.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018917.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018917.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018918.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018918.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018918.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018919.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018919.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018919.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018930.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018930.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018930.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018936.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018936.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018936.mm.1 NM_023229.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018922.mm.1 TC0500002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500018922.mm.1 NM_023229.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500018922.mm.1 NM_023229.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 918834 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0