Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029802.4 | NP_084078.3 | 5 - 6 | 746 | 100 | |
GenBank mRNA | AK154772.1 | - | 5 - 6 | 712 | 100 | |
BC013794.1 | - | 5 - 6 | 657 | 2648132 | 100 | |
BC022942.1 | - | 5 - 6 | 670 | 3583149 | 100 | |
BC028310.1 | - | 5 - 6 | 661 | 100 | ||
BC034520.1 | - | 5 - 6 | 727 | 3964827 | 100 | |
BC079603.1 | - | 2 - 3 | 1919 | 100 | ||
BU583325.1 | - | 1 - 2 | 201 | 100 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |