Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01172201_g1
          See other 1700037J18RIK GE Assays ›
          Gene Symbol:
          1700037J18Rik
          Gene Name:
          RIKEN cDNA 1700037J18 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01172201_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700037J18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006623.1 - 1 - 2 68 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY706620.1 - 1 - 2 67 Chr. 9 - 16599380 - 16599802 [+] on Build GRCm38 108

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream FAT tumor suppressor homolog 3 (Drosophila) Fat3 9430076A06Rik,
          D430038H04Rik,
          Gm1132,
          Gm510,
          MFAT3F
          -96.5kb Anti-sense Chr. 9 - 15910189 - 16502879 on NCBI Build GRCm38
          Downstream cysteine and histidine-rich domain (CHORD)-containing,
          zinc-binding protein 1
          Chordc1 1110001O09Rik,
          AA409036,
          Chp-1,
          morgana
          1.7mb Sense Chr. 9 - 18292233 - 18314000 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08002874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07689121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07996633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07455424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07455436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26329906 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 588749 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0