Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cactin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027381.2 | NP_081657.1 | 4 - 5 | 1223 | 65 | |
GenBank mRNA | AY934504.1 | - | 4 - 5 | 1129 | 65 | |
BC023696.1 | - | 1 - 2 | 143 | 65 | ||
BC150986.1 | - | 4 - 5 | 1223 | 9087899 | 65 | |
BC151180.1 | - | 4 - 5 | 1223 | 9088069 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |