Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01174180_m1
          See other PML GE Assays ›
          Gene Symbol:
          Pml
          Gene Name:
          promyelocytic leukemia
          Gene Aliases:
          1200009E24Rik, AI661194, Trim19
          Chromosome Location:
          Chr.9: 58217179 - 58249796 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01174180_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pml
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008884.5 NP_032910.3 3 - 4 1293 89
          NM_178087.4 NP_835188.2 3 - 4 1293 89
          GenBank mRNA AK032799.1 - 1 - 2 173 89
          AK052305.1 - 3 - 4 1293 89
          AK054135.1 - 3 - 4 1292 89
          AK137538.1 - 3 - 4 1241 89
          AK156279.1 - 3 - 4 1175 89
          AK192347.1 - 207 89
          BC020990.1 - 3 - 4 1238 4188386 89
          KJ650238.1 - 3 - 4 1197 89
          U33626.2 - 3 - 4 1190 89
          U89412.1 - 1 - 2 278 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002389.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pml

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of protein phosphorylation
          positive regulation of defense response to virus by host
          immune system process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein targeting
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          cell aging
          negative regulation of cell population proliferation
          intrinsic apoptotic signaling pathway in response to DNA damage
          intrinsic apoptotic signaling pathway in response to oxidative stress
          response to UV
          response to gamma radiation
          regulation of calcium ion transport into cytosol
          viral process
          negative regulation of angiogenesis
          myeloid cell differentiation
          negative regulation of cell growth
          PML body organization
          positive regulation of histone deacetylation
          negative regulation of telomere maintenance via telomerase
          endoplasmic reticulum calcium ion homeostasis
          negative regulation of interleukin-1 production
          circadian regulation of gene expression
          negative regulation of translation in response to oxidative stress
          response to cytokine
          regulation of circadian rhythm
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          entrainment of circadian clock by photoperiod
          proteasome-mediated ubiquitin-dependent protein catabolic process
          innate immune response
          cell fate commitment
          regulation of MHC class I biosynthetic process
          negative regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          retinoic acid receptor signaling pathway
          rhythmic process
          protein stabilization
          maintenance of protein location in nucleus
          defense response to virus
          negative regulation of telomerase activity
          positive regulation of apoptotic process involved in mammary gland involution
          regulation of SMAD protein signal transduction
          branching involved in mammary gland duct morphogenesis
          protein-containing complex assembly
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to interleukin-4
          intrinsic apoptotic signaling pathway by p53 class mediator
          cellular senescence
          extrinsic apoptotic signaling pathway
          negative regulation of viral release from host cell
          regulation of nucleic acid-templated transcription
          negative regulation of ubiquitin-dependent protein catabolic process
          regulation of double-strand break repair
          positive regulation of apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway
          DNA binding
          transcription coactivator activity
          protein binding
          zinc ion binding
          ubiquitin protein ligase binding
          SUMO binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          protein heterodimerization activity
          cobalt ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900020088.mm.1 TC0900002389.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020088.mm.1 NM_008884.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020088.mm.1 NM_008884.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020088.mm.1 NM_178087.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020088.mm.1 NM_178087.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.131.9:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0