Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175412_m1
          See other GM20745 GE Assays ›
          Gene Symbol:
          Gm20745
          Gene Name:
          predicted gene, 20745
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 68017964 - 68025714 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175412_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm20745
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045745.1 2 - 3 700 102
          GenBank mRNA AK045659.1 - 2 - 3 701 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream vacuolar protein sorting 13C (yeast) Vps13c AI552542,
          AV026040,
          C230055H22Rik,
          W51662,
          chorein
          -22.3kb Anti-sense Chr. 9 - 67840331 - 67995640 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930502A04 gene 4930502A04Rik 445.9kb Sense Chr. 9 - 68470902 - 68520200 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002566.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165582 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08095654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08175153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07728501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07681395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07474969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07548734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08090029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07572090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07728507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08175158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07728508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26602414 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900021472.mm.1 TC0900002566.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900021472.mm.1 NR_045745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021472.mm.1 NR_045745.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 515032 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0