Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175433_g1
          See other SEPT5 GE Assays ›
          Gene Symbol:
          Sept5
          Gene Name:
          septin 5
          Gene Aliases:
          Cdcrel-1, Cdcrel1, Pnutl1
          Chromosome Location:
          Chr.16: 18621811 - 18629938 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175433_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sept5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_213614.2 NP_998779.2 11 - 12 1142 74
          GenBank mRNA AK134700.1 - 11 - 12 1079 74
          BC059848.1 - 9 - 10 1020 74
          BC079631.1 - 5 - 6 456 74
          BC094347.1 - 10 - 11 1018 74
          BC141073.1 - 11 - 12 1152 9055685 74
          BC145331.1 - 11 - 12 1152 9053887 74
          BC145332.1 - 11 - 12 1201 9053888 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001319.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sept5

          Gene Ontology Categories:

          Function(s) Process(es)

          cell cycle
          regulation of exocytosis
          positive regulation of exocytosis
          cell division
          nucleotide binding
          protein binding
          GTP binding
          GDP binding
          syntaxin binding
          phosphatidylinositol binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC094347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC141073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC145331.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC145332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039881 BC059848.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206384 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554976 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC094347.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC141073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC145331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC145332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039881 BC059848.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 BC094347.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 BC059848.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 AK134700.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 BC079631.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P464530 NM_213614.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC094347.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC141073.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC145331.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC145332.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P21353 BC059848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC094347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC141073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC145331.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC145332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P21353 BC059848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 BC094347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 BC059848.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 AK134700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 BC079631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464530 NM_213614.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637534 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637536 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637553 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12637542 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438365 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438367 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438368 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438364 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438366 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438370 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438369 - -
          Affymetrix Mouse Gene 1.1 ST Array 10438358 - -
          Mouse Genome 430 2.0 Array 1452357_PM_at - -
          Mouse Genome 430 2.0 Array 1452357_at - -
          Affymetrix Mouse Clariom S Array PSR1600010878.mm.1 TC1600001319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010878.mm.1 NM_001001999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010878.mm.1 NM_001001999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010878.mm.1 NM_010327.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010878.mm.1 NM_010327.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010878.mm.1 NM_213614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010878.mm.1 NM_213614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010879.mm.1 TC1600001319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010879.mm.1 NM_213614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010879.mm.1 NM_213614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010877.mm.1 TC1600001319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010877.mm.1 NM_001001999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010877.mm.1 NM_001001999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010877.mm.1 NM_010327.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010877.mm.1 NM_010327.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010877.mm.1 NM_213614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010877.mm.1 NM_213614.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010880.mm.1 TC1600001319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010880.mm.1 NM_213614.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010880.mm.1 NM_213614.2 Refseq
          Mouse Expression Set 430 1452357_PM_at - -
          Mouse Expression Set 430 1452357_at - -
          Murine Genome U74v2 Set 96337_PM_at - -
          Murine Genome U74v2 Set 96337_at - -
          Mouse Genome 430A 2.0 Array 1452357_PM_at - -
          Mouse Genome 430A 2.0 Array 1452357_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 836508 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2828265 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2828265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0