Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01175451_m1
          See other GM20611 GE Assays ›
          Gene Symbol:
          Gm20611
          Gene Name:
          predicted gene 20611
          Gene Aliases:
          4930565A17
          Chromosome Location:
          Chr.10: 61031303 - 61038080 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01175451_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm20611
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040638.1 2 - 3 928 67
          GenBank mRNA AK029795.1 - 2 - 3 928 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB016902.2 - 1 - 2 487 Chr. 10 - 61036723 - 61038080 [+] on Build GRCm38 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          26947
          Gm26947 -95.5kb Sense Chr. 10 - 60931986 - 60940944 on NCBI Build GRCm38
          Downstream pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 1 Pcbd1 Dcoh,
          Pcbd,
          Pcd,
          Phs
          51.3kb Sense Chr. 10 - 61089331 - 61094329 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000557.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2413727 AK029795.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222805 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155783 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155783 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100196 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08691341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08481659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08278713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08293771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08966828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08690312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08278715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08723811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08690311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08627148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08516607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08482531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08423094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08944362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08482556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08944360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08691346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08966827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08227648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08944355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08967501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08944365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08691365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08966825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08348028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08691347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08586875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08967490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27178169 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1446251_PM_at - -
          Mouse Genome 430 2.0 Array 1446251_PM_at - -
          Mouse Genome 430 2.0 Array 1446251_at - -
          Affymetrix Mouse Clariom S Array PSR1000003780.mm.1 TC1000000557.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000003780.mm.1 NR_040638.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003780.mm.1 NR_040638.1 Refseq
          Mouse Expression Set 430 1446251_PM_at - -
          Mouse Expression Set 430 1446251_at - -
          Affymetrix HT Mouse Genome 430B Array 1446251_at - -
          Affymetrix HT Mouse Genome 430B Array 1446251_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 846211 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104200333 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix