Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176957_g1
          See other COX5A GE Assays ›
          Gene Symbol:
          Cox5a
          Gene Name:
          cytochrome c oxidase subunit Va
          Gene Aliases:
          AA959768, CcOX
          Chromosome Location:
          Chr.9: 57521232 - 57532427 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176957_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007747.2 NP_031773.2 3 - 4 422 91
          GenBank mRNA AK018723.1 - 3 - 4 357 91
          BC003831.1 - 2 - 3 1548 91
          BC034302.1 - 3 - 4 375 5372849 91
          CO044753.1 - 2 - 3 91
          X15963.1 - 3 - 4 346 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000705.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox5a

          Gene Ontology Categories:

          Function(s) Process(es)

          proton transmembrane transport
          cytochrome-c oxidase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542211 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542217 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335900 X15963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335900 X15963.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347273 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764859 NM_146503.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764859 NM_207141.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64292 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118383 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141798 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335900 X15963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335900 X15963.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P335900 X15963.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P335900 X15963.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26542214 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141798 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06801898 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118383 - -
          Whole Mouse Genome G4122A A_52_P121110 - -
          Whole Mouse Genome G4122A A_51_P335900 X15963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335900 X15963.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121497 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118383 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08174155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08174151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07495289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08174141 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542211 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26542214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26542197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08032972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08174142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07698837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07956983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08184409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07771177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08155022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08174151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08174140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07597222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26542248 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10585711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585712 - -
          Affymetrix HT MG-430 PM Array Plate 1439267_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1415933_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1448153_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10585706 - -
          Mouse Genome 430 2.0 Array 1415933_PM_a_at - -
          Mouse Genome 430 2.0 Array 1439267_x_at - -
          Mouse Genome 430 2.0 Array 1448153_PM_at - -
          Mouse Genome 430 2.0 Array 1439267_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448153_at - -
          Mouse Genome 430 2.0 Array 1415933_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12858_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12858_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900005184.mm.1 TC0900000705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900005184.mm.1 NM_007747.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900005184.mm.1 NM_007747.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900005185.mm.1 TC0900000705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900005185.mm.1 NM_007747.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900005185.mm.1 NM_007747.2 Refseq
          Mouse Expression Set 430 1448153_at - -
          Mouse Expression Set 430 1439267_PM_x_at - -
          Mouse Expression Set 430 1415933_a_at - -
          Mouse Expression Set 430 1415933_PM_a_at - -
          Mouse Expression Set 430 1439267_x_at - -
          Mouse Expression Set 430 1448153_PM_at - -
          Murine Genome U74v2 Set 161490_PM_at - -
          Murine Genome U74v2 Set 160438_at - -
          Murine Genome U74v2 Set 160438_PM_at - -
          Murine Genome U74v2 Set 161490_at - -
          Murine Genome U74v2 Set 171593_PM_at - -
          Murine Genome U74v2 Set 171593_at - -
          Mouse Genome 430A 2.0 Array 1439267_x_at - -
          Mouse Genome 430A 2.0 Array 1439267_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1448153_at - -
          Mouse Genome 430A 2.0 Array 1415933_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1415933_a_at - -
          Mouse Genome 430A 2.0 Array 1448153_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439267_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415933_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448153_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448153_at - -
          Affymetrix HT Mouse Genome 430A Array 1439267_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415933_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893012 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1450609 - -
          Illumina mouse-6 v1.0 expression beadchip 1450609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2682800 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2682800 - -
          mouse-6 v1.1 (Illumina) scl36904.3.10_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36904.3.10_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450609 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2682800 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2682800 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36904.3.10_19-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7ffdcc7b59-b9k6r:80/100.66.134.75:80.
          git-commit: c6f4609997b4833aba2fd3ad806738f0433cd95c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc