Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01177546_mH
          See other GSTT3 GE Assays ›
          Gene Symbol:
          Gstt3
          Gene Name:
          glutathione S-transferase, theta 3
          Gene Aliases:
          AI118089
          Chromosome Location:
          Chr.10: 75774122 - 75781483 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01177546_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gstt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133994.3 NP_598755.1 3 - 4 812 54
          GenBank mRNA AF508157.1 - 3 - 4 812 54
          AK138669.1 - 3 - 4 469 54
          AK154161.1 - 3 - 4 518 54
          AK168226.1 - 3 - 4 461 54
          BC003903.1 - 3 - 4 440 3601446 54
          BC057964.1 - 3 - 4 767 5352325 54
          BC062201.1 - 3 - 4 769 4913425 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003226.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gstt3

          Gene Ontology Categories:

          Function(s) Process(es)

          glutathione metabolic process
          glutathione transferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 BC003903.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 BC057964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 BC062201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 AK138669.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 AK154161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 BC003903.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 BC057964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 BC062201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 AK138669.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377856 AK154161.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 523333 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 758290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114822 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108639 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 BC003903.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 BC057964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 BC062201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 AK138669.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 AK154161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 BC003903.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 BC057964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 BC062201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 AK138669.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377856 AK154161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 BC003903.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 BC057964.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 BC062201.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 AK138669.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 AK154161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 BC003903.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 BC057964.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 BC062201.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 AK138669.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P377856 AK154161.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27258626 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07111248 - -
          Whole Mouse Genome G4122A A_51_P377856 BC003903.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 BC057964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 BC062201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 AK138669.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 AK154161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 BC003903.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 BC057964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 BC062201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 AK138669.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377856 AK154161.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115505 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108639 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08918672 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08918681 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08918669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08417365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08918674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08803819 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08918670 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08918668 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08417363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27258533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08517824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08495665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08918669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08727291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08364620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08494322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08587164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08364618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08364614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27258622 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10369995 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369999 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369997 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369996 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369994 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369998 - -
          Affymetrix HT MG-430 PM Array Plate 1423891_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10369993 - -
          Mouse Genome 430 2.0 Array 1423891_at - -
          Mouse Genome 430 2.0 Array 1423891_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 103140_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 103140_at - -
          Affymetrix Mouse Clariom S Array PSR1000017037.mm.1 TC1000003226.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017037.mm.1 NM_133994.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017037.mm.1 NM_133994.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017041.mm.1 TC1000003226.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017041.mm.1 NM_133994.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017041.mm.1 NM_133994.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017036.mm.1 TC1000003226.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017036.mm.1 NM_133994.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017036.mm.1 NM_133994.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017035.mm.1 TC1000003226.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017035.mm.1 NM_133994.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017035.mm.1 NM_133994.3 Refseq
          Mouse Expression Set 430 1423891_at - -
          Mouse Expression Set 430 1423891_PM_at - -
          Mouse Genome 430A 2.0 Array 1423891_at - -
          Mouse Genome 430A 2.0 Array 1423891_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423891_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423891_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5080286 - -
          Illumina mouse-6 v1.0 expression beadchip 5080286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2665715 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2665715 - -
          mouse-6 v1.1 (Illumina) scl37812.5_586-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37812.5_586-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2665715 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2665715 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37812.5_586-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0