Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01178297_m1
          See other CCPG1OS GE Assays ›
          Gene Symbol:
          Ccpg1os
          Gene Name:
          cell cycle progression 1, opposite strand
          Gene Aliases:
          EG546143, Gm5918
          Chromosome Location:
          Chr.9: 72979717 - 72985376 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01178297_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccpg1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001198789.1 NP_001185718.1 1 - 2 232 77
          GenBank mRNA BC048630.1 - 1 - 2 215 77
          BC099568.1 - 1 - 2 209 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002651.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_001198789.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56595 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P520139 BC099568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P520139 AW125236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P520139 BC048630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P520139 NM_001198789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P520139 BC099568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P520139 AW125236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P520139 BC048630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P520139 NM_001198789.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01970087 - -
          Whole Mouse Genome G4122A A_51_P520139 BC099568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P520139 AW125236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P520139 BC048630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P520139 NM_001198789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P520139 BC099568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P520139 AW125236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P520139 BC048630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P520139 NM_001198789.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1452935_PM_at - -
          Mouse Genome 430 2.0 Array 1452935_at - -
          Mouse Genome 430 2.0 Array 1452935_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 546143_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 546143_at - -
          Affymetrix Mouse Clariom S Array PSR0900021989.mm.1 TC0900002651.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900021989.mm.1 NM_001198789.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021989.mm.1 NM_001198789.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021991.mm.1 TC0900002651.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900021991.mm.1 NM_001198789.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900021991.mm.1 NM_001198789.1 Refseq
          Affymetrix HT Mouse Genome 430B Array 1452935_at - -
          Affymetrix HT Mouse Genome 430B Array 1452935_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 577112 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257755 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0