Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01178819_m1
          See other TGFB1 GE Assays ›
          Gene Symbol:
          Tgfb1
          Gene Name:
          transforming growth factor, beta 1
          Gene Aliases:
          TGF-beta1, TGFbeta1, Tgfb, Tgfb-1
          Chromosome Location:
          Chr.7: 25687002 - 25704996 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01178819_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011577.1 NP_035707.1 2 - 3 1373 135
          GenBank mRNA AJ009862.1 - 2 - 3 1373 135
          AK144163.1 - 2 - 3 1375 135
          BC013738.1 - 2 - 3 820 3586216 135
          M13177.1 - 2 - 3 858 135

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000424.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgfb1

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          skeletal system development
          morphogenesis of a branching structure
          cell activation
          epithelial to mesenchymal transition
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          regulation of sodium ion transport
          chondrocyte differentiation
          hematopoietic progenitor cell differentiation
          CD4-positive, CD25-positive, alpha-beta regulatory T cell lineage commitment
          adaptive immune response based on somatic recombination of immune receptors built from immunoglobulin superfamily domains
          tolerance induction to self antigen
          protein phosphorylation
          protein export from nucleus
          ATP biosynthetic process
          phosphate-containing compound metabolic process
          cellular calcium ion homeostasis
          defense response
          inflammatory response
          cell cycle arrest
          mitotic cell cycle checkpoint
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          SMAD protein complex assembly
          SMAD protein import into nucleus
          Notch signaling pathway
          negative regulation of neuroblast proliferation
          endoderm development
          skeletal muscle tissue development
          negative regulation of DNA replication
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          germ cell migration
          defense response to fungus, incompatible interaction
          organ morphogenesis
          response to organic substance
          regulation of gene expression
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          positive regulation of fibroblast migration
          positive regulation of peptidyl-threonine phosphorylation
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of macrophage cytokine production
          cell growth
          regulation of striated muscle tissue development
          regulation of transforming growth factor beta receptor signaling pathway
          evasion or tolerance of host defenses by virus
          negative regulation of cell-cell adhesion
          hyaluronan catabolic process
          T cell differentiation
          negative regulation of ossification
          negative regulation of cell growth
          positive regulation of cell migration
          mammary gland development
          positive regulation of histone deacetylation
          organ regeneration
          positive regulation of protein complex assembly
          positive regulation of exit from mitosis
          lipopolysaccharide-mediated signaling pathway
          positive regulation of cellular protein metabolic process
          response to estradiol
          response to progesterone
          regulation of interleukin-23 production
          negative regulation of interleukin-17 production
          positive regulation of interleukin-17 production
          receptor catabolic process
          regulation of CD4-positive, CD25-positive, alpha-beta regulatory T cell differentiation
          positive regulation of superoxide anion generation
          mononuclear cell proliferation
          positive regulation of collagen biosynthetic process
          positive regulation of peptidyl-serine phosphorylation
          positive regulation of histone acetylation
          positive regulation of protein dephosphorylation
          growth
          wound healing
          T cell activation
          regulation of cell proliferation
          negative regulation of T cell proliferation
          regulation of protein import into nucleus
          positive regulation of protein import into nucleus
          positive regulation of odontogenesis
          myelination
          regulation of apoptotic process
          myeloid dendritic cell differentiation
          T cell homeostasis
          positive regulation of apoptotic process
          positive regulation of MAP kinase activity
          regulation of MAPK cascade
          protein kinase B signaling
          positive regulation of blood vessel endothelial cell migration
          negative regulation of blood vessel endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase activity
          regulatory T cell differentiation
          cell-cell junction organization
          negative regulation of fat cell differentiation
          negative regulation of myoblast differentiation
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          positive regulation of isotype switching to IgA isotypes
          cell development
          lymph node development
          negative regulation of skeletal muscle tissue development
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          positive regulation of protein secretion
          negative regulation of phagocytosis
          negative regulation of immune response
          negative regulation of T cell activation
          positive regulation of chemotaxis
          positive regulation of NF-kappaB transcription factor activity
          regulation of binding
          regulation of DNA binding
          positive regulation of smooth muscle cell differentiation
          negative regulation of release of sequestered calcium ion into cytosol
          positive regulation of cell division
          positive regulation of protein kinase B signaling
          pathway-restricted SMAD protein phosphorylation
          positive regulation of SMAD protein import into nucleus
          SMAD protein signal transduction
          mammary gland branching involved in thelarche
          branch elongation involved in mammary gland duct branching
          regulation of branching involved in mammary gland duct morphogenesis
          negative regulation of gene silencing by miRNA
          regulation of cartilage development
          lens fiber cell differentiation
          response to cholesterol
          positive regulation of cell cycle arrest
          cellular response to growth factor stimulus
          cellular response to organic cyclic compound
          cellular response to dexamethasone stimulus
          cellular response to transforming growth factor beta stimulus
          T-helper 17 cell lineage commitment
          extracellular matrix assembly
          positive regulation of branching involved in ureteric bud morphogenesis
          extrinsic apoptotic signaling pathway
          negative regulation of hyaluronan biosynthetic process
          positive regulation of protein localization to nucleus
          positive regulation of NAD+ ADP-ribosyltransferase activity
          regulation of actin cytoskeleton reorganization
          positive regulation of transcription regulatory region DNA binding
          glycoprotein binding
          antigen binding
          type II transforming growth factor beta receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity
          enzyme binding
          type I transforming growth factor beta receptor binding
          type III transforming growth factor beta receptor binding
          protein homodimerization activity
          protein heterodimerization activity
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700003313.mm.1 TC0700000424.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003313.mm.1 NM_011577.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0