Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179456_m1
          See other CD276 GE Assays ›
          Gene Symbol:
          Cd276
          Gene Name:
          CD276 antigen
          Gene Aliases:
          6030411F23Rik, AU016588, B7RP-2, B7h3
          Chromosome Location:
          Chr.9: 58524300 - 58555033 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179456_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd276
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133983.4 NP_598744.1 6 - 7 1137 63
          GenBank mRNA AK031354.1 - 6 - 7 868 63
          AK155114.1 - 7 - 8 1223 63
          AK204836.1 - 1 - 2 38 63
          AY190318.1 - 6 - 7 952 63
          BC019436.1 - 6 - 7 1041 3674228 63
          BC056608.1 - 6 - 7 1138 6397680 63
          DQ832276.1 - 7 - 8 1236 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002396.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd276

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of bone mineralization
          positive regulation of interleukin-2 production
          positive regulation of T cell proliferation
          T cell activation
          negative regulation of T cell proliferation
          positive regulation of osteoblast differentiation
          negative regulation of inflammatory response
          regulation of immune response
          signaling receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499551 BC019436.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P499551 BC019436.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 548597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81964 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179801 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102426 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108039 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499551 BC019436.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P499551 BC019436.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P499551 BC019436.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P499551 BC019436.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26548541 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06804569 - -
          Whole Mouse Genome G4122A A_51_P499551 BC019436.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P499551 BC019436.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116056 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136487 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06804569 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07762554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07762561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07762558 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07666230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07762556 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07762555 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07762560 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07762559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26548559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07668383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07496118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07482982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07457064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07860813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07666231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07541747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07762555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07484538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07784163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07457066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07496122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07457065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548565 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10594102 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585841 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594101 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10594094 - -
          Affymetrix HT MG-430 PM Array Plate 1417599_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10594092 - -
          Affymetrix Mouse Gene 1.1 ST Array 10585840 - -
          Mouse Genome 430 2.0 Array 1417599_at - -
          Mouse Genome 430 2.0 Array 1417599_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102657_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102657_at - -
          Affymetrix Mouse Clariom S Array PSR0900020142.mm.1 TC0900002396.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020142.mm.1 NM_133983.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020142.mm.1 NM_133983.4 Refseq
          Mouse Expression Set 430 1417599_PM_at - -
          Mouse Expression Set 430 1417599_at - -
          Murine Genome U74v2 Set 93174_PM_at - -
          Murine Genome U74v2 Set 93174_at - -
          Mouse Genome 430A 2.0 Array 1417599_at - -
          Mouse Genome 430A 2.0 Array 1417599_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417599_at - -
          Affymetrix HT Mouse Genome 430A Array 1417599_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 513587 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3840019 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259610 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259610 - -
          mouse-6 v1.1 (Illumina) scl0102657.1_90-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0102657.1_90-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840019 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1259610 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259610 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0