Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AU040320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035525.1 | NP_001030602.1 | 10 - 11 | 1786 | 54 | |
NM_001035526.1 | NP_001030603.1 | 10 - 11 | 1904 | 54 | ||
NM_133886.2 | NP_598647.1 | 10 - 11 | 1904 | 54 | ||
GenBank mRNA | AK031342.1 | - | 10 - 11 | 1848 | 54 | |
AK043006.1 | - | 10 - 11 | 1854 | 54 | ||
AK049570.1 | - | 10 - 11 | 1787 | 54 | ||
AK084668.1 | - | 10 - 11 | 1749 | 54 | ||
AK147569.1 | - | 4435 | 54 | |||
AK170261.1 | - | 10 - 11 | 1861 | 54 | ||
AK179638.1 | - | 1 - 2 | 201 | 54 | ||
AK208774.1 | - | 2 - 3 | 346 | 54 | ||
AK215400.1 | - | 1 - 2 | 200 | 54 | ||
BC022154.1 | - | 1 - 2 | 316 | 54 | ||
BC028869.1 | - | 10 - 11 | 1874 | 4235813 | 54 | |
CA540237.1 | - | 1 - 2 | 210 | 54 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |