Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AU040320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035525.1 | NP_001030602.1 | 8 - 9 | 1430 | 84 | |
NM_001035526.1 | NP_001030603.1 | 8 - 9 | 1548 | 84 | ||
NM_133886.2 | NP_598647.1 | 8 - 9 | 1548 | 84 | ||
GenBank mRNA | AK031342.1 | - | 8 - 9 | 1492 | 84 | |
AK043006.1 | - | 8 - 9 | 1498 | 84 | ||
AK049570.1 | - | 8 - 9 | 1431 | 84 | ||
AK084668.1 | - | 8 - 9 | 1393 | 84 | ||
AK147569.1 | - | 4079 | 84 | |||
AK170261.1 | - | 8 - 9 | 1505 | 84 | ||
AK190417.1 | - | 1 - 2 | 82 | 84 | ||
AK191074.1 | - | 1 - 2 | 82 | 84 | ||
BC028869.1 | - | 8 - 9 | 1518 | 4235813 | 84 | |
CB527694.1 | - | 4 - 5 | 350 | 84 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |