Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01179861_m1
          See other CEP85L GE Assays ›
          Gene Symbol:
          Cep85l
          Gene Name:
          centrosomal protein 85-like
          Gene Aliases:
          ENSMUSG00000038594, Gm9766
          Chromosome Location:
          Chr.10: 53276799 - 53380701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01179861_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cep85l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204983.1 NP_001191912.1 12 - 13 2528 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002164.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115161 NM_001204983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115161 AK039734.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440416 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464739 AK156593.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526798 AK136569.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526798 NM_001033444.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 221807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142316 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115161 NM_001204983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115161 AK039734.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02126047 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02126012 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061090 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061108 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061159 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061271 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061260 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061134 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07061146 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07061271 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07061134 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07061225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27142001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08558597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27142005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27142004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08845937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08845957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08680977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27142003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27142009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141687 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10369114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369115 - -
          Affymetrix Mouse Clariom S Array PSR1000015162.mm.1 TC1000002164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015162.mm.1 NM_001204983.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015162.mm.1 NM_001204983.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015165.mm.1 TC1000002164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000015165.mm.1 NM_001204983.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015165.mm.1 NM_001204983.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 765379 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0