Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028006.2 | NP_082282.1 | 7 - 8 | 691 | 97 | |
GenBank mRNA | AK010005.1 | - | 7 - 8 | 685 | 97 | |
AK041416.1 | - | 7 - 8 | 656 | 97 | ||
AK148039.1 | - | 7 - 8 | 691 | 97 | ||
BC062179.1 | - | 7 - 8 | 688 | 6333817 | 97 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |