Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400009502.mm.1 |
TC0400001269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400009502.mm.1 |
NM_001029912.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400009502.mm.1 |
NM_001029912.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400009501.mm.1 |
TC0400001269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400009501.mm.1 |
NM_001029912.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400009501.mm.1 |
NM_001029912.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400009500.mm.1 |
TC0400001269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400009500.mm.1 |
NM_001029912.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400009500.mm.1 |
NM_001029912.2 |
Refseq |