Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01182623_g1
          See other PRADC1 GE Assays ›
          Gene Symbol:
          Pradc1
          Gene Name:
          protease-associated domain containing 1
          Gene Aliases:
          1700040I03Rik, Pap21
          Chromosome Location:
          Chr.6: 85446785 - 85451970 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01182623_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pradc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163427.1 NP_001156899.1 4 - 5 566 70
          NR_028081.1 4 - 5 590 70
          GenBank mRNA AK006658.1 - 4 - 5 532 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002613.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605100 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605097 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605092 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605095 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605098 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605093 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605099 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605101 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605094 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605096 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040416 NR_028081.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040416 AK006658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040416 BC115452.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040416 NM_028505.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2040416 CJ208123.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45835 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128343 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040416 NR_028081.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040416 AK006658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040416 BC115452.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040416 NM_028505.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2040416 CJ208123.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128343 - -
          Whole Mouse Genome G4122A A_51_P454447 NR_028081.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454447 AK006658.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454447 BC115452.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454447 NM_028505.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P454447 CJ208123.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05419025 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05419011 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05419015 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605099 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605101 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605092 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605096 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605097 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605094 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605100 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605093 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605095 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24605098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24604998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24605012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05288574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05331201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05681617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05288557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24605066 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545839 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545841 - -
          Affymetrix HT MG-430 PM Array Plate 1429122_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545835 - -
          Mouse Genome 430 2.0 Array 1429122_a_at - -
          Mouse Genome 430 2.0 Array 1429122_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73327_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73327_at - -
          Affymetrix Mouse Clariom S Array PSR0600020274.mm.1 TC0600002613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020274.mm.1 NM_001163427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020274.mm.1 NM_001163427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020274.mm.1 NM_028505.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020274.mm.1 NM_028505.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020274.mm.1 NR_028081.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020274.mm.1 NR_028081.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020277.mm.1 TC0600002613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020277.mm.1 NM_001163427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020277.mm.1 NM_001163427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020277.mm.1 NM_028505.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020277.mm.1 NM_028505.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020277.mm.1 NR_028081.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020277.mm.1 NR_028081.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020276.mm.1 TC0600002613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020276.mm.1 NM_001163427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020276.mm.1 NM_001163427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020276.mm.1 NM_028505.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020276.mm.1 NM_028505.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020276.mm.1 NR_028081.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020276.mm.1 NR_028081.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020273.mm.1 TC0600002613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020273.mm.1 NM_001163427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020273.mm.1 NM_001163427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020273.mm.1 NM_028505.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020273.mm.1 NM_028505.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020273.mm.1 NR_028081.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020273.mm.1 NR_028081.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020272.mm.1 TC0600002613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020272.mm.1 NM_001163427.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020272.mm.1 NM_001163427.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020272.mm.1 NM_028505.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020272.mm.1 NM_028505.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020272.mm.1 NR_028081.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020272.mm.1 NR_028081.1 Refseq
          Mouse Expression Set 430 1429122_PM_a_at - -
          Mouse Expression Set 430 1429122_a_at - -
          Murine Genome U74v2 Set 95619_PM_at - -
          Murine Genome U74v2 Set 95619_at - -
          Mouse Genome 430A 2.0 Array 1429122_a_at - -
          Mouse Genome 430A 2.0 Array 1429122_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429122_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429122_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 682782 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6450750 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2881024 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2881024 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-595d89cdf5-c6dj8:80/100.66.135.190:80.
          git-commit: bccefdb7e3170cc2d597cd09baf15c702f2b69d9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0