Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600022649.mm.1 |
TC0600002946.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022649.mm.1 |
NM_026894.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022649.mm.1 |
NM_026894.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022651.mm.1 |
TC0600002946.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022651.mm.1 |
NM_026894.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022651.mm.1 |
NM_026894.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022650.mm.1 |
TC0600002946.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022650.mm.1 |
NM_026894.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022650.mm.1 |
NM_026894.1 |
Refseq |