Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01183729_g1
          See other RAB7 GE Assays ›
          Gene Symbol:
          Rab7
          Gene Name:
          RAB7, member RAS oncogene family
          Gene Aliases:
          Rab7a
          Chromosome Location:
          Chr.6: 87999106 - 88045270 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01183729_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rab7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293652.1 NP_001280581.1 4 - 5 479 105
          NM_001293653.1 NP_001280582.1 2 - 3 140 105
          NM_001293654.1 NP_001280583.1 3 - 4 306 105
          NM_001293655.1 NP_001280584.1 2 - 3 219 105
          NM_009005.3 NP_033031.2 2 - 3 257 105
          GenBank mRNA AB232599.1 - 1 - 2 51 105
          AK005004.1 - 2 - 3 219 105
          AK057671.1 - 2 - 3 186 105
          AK135659.1 - 3 - 4 307 105
          AK139914.1 - 2 - 3 210 105
          AK148117.1 - 2 - 3 250 105
          AK151078.1 - 2 - 3 231 105
          AK152666.1 - 4 - 5 454 105
          AK154234.1 - 2 - 3 238 105
          AK158971.1 - 2 - 3 141 105
          AK159507.1 - 2 - 3 187 105
          AK159928.1 - 2 - 3 158 105
          AK179526.1 - 2 - 3 96 105
          AK190348.1 - 2 - 3 96 105
          BC004597.1 - 2 - 3 175 3584891 105
          BC086793.1 - 2 - 3 173 6414012 105
          BY291181.1 - 2 - 3 258 105
          CT010352.1 - 1 - 2 51 105
          X89650.1 - 2 - 3 207 105

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rab7

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to lysosome
          transport
          intracellular protein transport
          epidermal growth factor catabolic process
          small GTPase mediated signal transduction
          endosome to lysosome transport
          protein transport
          viral release from host cell
          protein to membrane docking
          Rab protein signal transduction
          retrograde transport, endosome to Golgi
          early endosome to late endosome transport
          bone resorption
          positive regulation of protein catabolic process
          positive regulation of viral process
          phagosome acidification
          phagosome-lysosome fusion
          positive regulation of exosomal secretion
          regulation of autophagosome assembly
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          GDP binding
          small GTPase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600020778.mm.1 TC0600002674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020778.mm.1 NM_009005.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020777.mm.1 TC0600002674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020777.mm.1 NM_009005.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020779.mm.1 TC0600002674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020779.mm.1 NM_009005.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020786.mm.1 TC0600002674.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 671756 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0