Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400013048.mm.1 |
TC0400001639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013048.mm.1 |
NM_019641.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013048.mm.1 |
NM_019641.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013034.mm.1 |
TC0400001639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013034.mm.1 |
NM_019641.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013034.mm.1 |
NM_019641.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013035.mm.1 |
TC0400001639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013035.mm.1 |
NM_019641.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013035.mm.1 |
NM_019641.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013045.mm.1 |
TC0400001639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013045.mm.1 |
NM_019641.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013045.mm.1 |
NM_019641.4 |
Refseq |