Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01186497_m1
          See other GM5577 GE Assays ›
          Gene Symbol:
          Gm5577
          Gene Name:
          predicted gene 5577
          Gene Aliases:
          EG434064
          Chromosome Location:
          Chr.6: 87981683 - 87984180 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01186497_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5577
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026990.1 1 - 2 202 112
          GenBank mRNA AK020089.1 - 204 112
          AK048951.1 - 1 - 2 174 112
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB650272.1 - 1 - 2 174 Chr. 6 - 87981712 - 87983439 [+] on Build GRCm38 112

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream H1 histone family,
          member X
          H1fx Gm461,
          H1.10,
          H1X
          -230.0 Anti-sense Chr. 6 - 87980421 - 87981482 on NCBI Build GRCm38
          Downstream RAB7,
          member RAS oncogene family
          Rab7 Rab7a 15.7kb Anti-sense Chr. 6 - 87999106 - 88045270 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001022.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386829 AK042699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386829 AK048951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386829 NR_026990.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386829 AK042699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386829 AK048951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386829 NR_026990.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 752750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138772 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143388 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176031 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386829 AK042699.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386829 AK048951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386829 NR_026990.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386829 AK042699.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386829 AK048951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386829 NR_026990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386829 AK042699.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386829 AK048951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386829 NR_026990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386829 AK042699.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386829 AK048951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386829 NR_026990.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539860 - -
          Affymetrix Mouse Gene 1.1 ST Array 10539857 - -
          Affymetrix Mouse Clariom S Array PSR0600008024.mm.1 TC0600001022.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008024.mm.1 NR_026990.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008024.mm.1 NR_026990.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008023.mm.1 TC0600001022.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008023.mm.1 NR_026990.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008023.mm.1 NR_026990.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008025.mm.1 TC0600001022.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600008025.mm.1 NR_026990.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600008025.mm.1 NR_026990.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 741369 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 105860102 - -
          Illumina Sentrix mouse-6.v1 bead array 105860102 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105860102 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257810 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix