Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01188570_g1
          See other PPIH GE Assays ›
          Gene Symbol:
          Ppih Gm10069
          Gene Name:
          peptidyl prolyl isomerase H
          predicted gene 10069
          Gene Aliases:
          1100001J08Rik, 2010111B15Rik, 4833408F11Rik, AI464484, CYP-20, CYPH, D4Wsu43e
          ENSMUSG00000059659
          Chromosome Location:
          Chr.4: 119300010 - 119320523 on Build GRCm38
          Chromosome Location:
          Chr.6: 128438757 - 128503281 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01188570_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppih
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110129.1 NP_001103599.1 8 - 9 521 71
          NM_028677.4 NP_082953.1 8 - 9 521 71
          GenBank mRNA AK003179.1 - 8 - 9 499 71
          AK005202.1 - 8 - 9 482 71
          AK014665.1 - 8 - 9 482 71
          AK134995.1 - 8 - 9 501 71
          AK136330.1 - 8 - 9 480 71
          AK164299.1 - 8 - 9 486 71
          BC016565.1 - 8 - 9 476 71
          BC050116.1 - 8 - 9 475 6432881 71
          BC098490.1 - 477 30022355 71
          Gene Symbol: Gm10069
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028592.1 3 71
          GenBank mRNA AK029950.1 - 3 71
          AK046088.1 - 3 71
          AK086937.1 - 3 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003422.mm.1, TC0600001510.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK046088.1, AK086937.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppih
          Click here to find siRNAs for Gene Gm10069

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          mRNA processing
          protein folding
          RNA splicing
          positive regulation of viral genome replication
          biological_process
          peptidyl-prolyl cis-trans isomerase activity
          isomerase activity
          ribonucleoprotein complex binding
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400027747.mm.1 TC0400003422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027747.mm.1 NM_028677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027747.mm.1 NM_028677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027755.mm.1 TC0400003422.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027755.mm.1 NM_001110129.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027755.mm.1 NM_001110129.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027755.mm.1 NM_028677.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027755.mm.1 NM_028677.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012338.mm.1 TC0600001510.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012338.mm.1 NR_028592.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012338.mm.1 NR_028592.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0