Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173002.3 | NP_766590.1 | 7 - 8 | 2382 | 95 | |
GenBank mRNA | AK044429.1 | - | 7 - 8 | 2275 | 95 | |
AK047079.1 | - | 7 - 8 | 2264 | 95 | ||
AK047538.1 | - | 7 - 8 | 2298 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() |