Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01189933_g1
          See other CENPW GE Assays ›
          Gene Symbol:
          Cenpw
          Gene Name:
          centromere protein W
          Gene Aliases:
          2610036L11Rik, Cug2
          Chromosome Location:
          Chr.10: 30196009 - 30200600 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01189933_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cenpw
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109747.1 NP_001103217.1 2 - 3 289 108
          GenBank mRNA AK005988.1 - 2 - 3 271 108
          AK011700.1 - 2 - 3 270 107
          AK141272.1 - 2 - 3 271 108
          BC147188.1 - 2 - 3 286 9056212 108
          BC147189.1 - 2 - 3 286 9056237 108
          BC147710.1 - 2 - 3 286 9087469 108
          BC147721.1 - 2 - 3 286 9087480 108
          BG804382.1 - 2 - 3 289 108

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          cell cycle
          chromosome segregation
          chromosome organization
          cell division
          kinetochore assembly
          molecular_function
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 BC147721.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 BG804382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 AK141272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 BC050071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 BC147188.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 BC147721.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 BG804382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 AK141272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 BC050071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P609024 BC147188.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175171 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 BC147721.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 BG804382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 AK141272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 BC050071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 BC147188.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 BC147721.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 BG804382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 AK141272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 BC050071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P609024 BC147188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P126070 BC147721.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P126070 AK011700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P126070 AK141272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P126070 BC050071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P126070 BC147188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 BC147721.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 BG804382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 AK141272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 BC050071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 BC147188.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 BC147721.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 BG804382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 AK141272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 BC050071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609024 BC147188.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27038651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08800864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08711624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08536768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08536757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08800861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08711627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08528906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27038654 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10368510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368509 - -
          Affymetrix HT MG-430 PM Array Plate 1429156_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453053_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10368508 - -
          Mouse Genome 430 2.0 Array 1429156_PM_at - -
          Mouse Genome 430 2.0 Array 1429156_at - -
          Mouse Genome 430 2.0 Array 1453053_at - -
          Mouse Genome 430 2.0 Array 1453053_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66311_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66311_at - -
          Affymetrix Mouse Clariom S Array PSR1000013956.mm.1 TC1000001941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013956.mm.1 NM_001109747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013956.mm.1 NM_001109747.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013958.mm.1 TC1000001941.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013958.mm.1 NM_001109747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013958.mm.1 NM_001109747.1 Refseq
          Mouse Expression Set 430 1429156_PM_at - -
          Mouse Expression Set 430 1429156_at - -
          Mouse Expression Set 430 1453053_at - -
          Mouse Expression Set 430 1453053_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429156_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453053_at - -
          Affymetrix HT Mouse Genome 430B Array 1429156_at - -
          Affymetrix HT Mouse Genome 430B Array 1453053_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 611038 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106370692 - -
          Illumina mouse-6 v1.0 expression beadchip 106370692 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2543929 - -
          Illumina Sentrix mouse-6.v1 bead array 106370692 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106370692 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2543929 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0