Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01190763_m1
          See other CATSPER4 GE Assays ›
          Gene Symbol:
          Catsper4
          Gene Name:
          cation channel, sperm associated 4
          Gene Aliases:
          4933412L05
          Chromosome Location:
          Chr.4: 134211968 - 134227434 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01190763_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Catsper4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130030.1 NP_001123502.1 8 - 9 1106 124
          NM_177866.4 NP_808534.1 10 - 11 1325 124
          GenBank mRNA AK077145.1 - 10 - 11 1312 124
          BC108974.2 - 10 - 11 1305 40047424 124
          BC108975.1 - 10 - 11 1305 40047425 124
          EU781510.1 - 9 - 10 1202 124
          EU781511.1 - 8 - 9 1081 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003730.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Catsper4

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          sodium ion transport
          calcium ion transport
          multicellular organism development
          spermatogenesis
          cell differentiation
          flagellated sperm motility
          regulation of ion transmembrane transport
          sperm capacitation
          transmembrane transport
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during action potential
          ion channel activity
          calcium activated cation channel activity
          voltage-gated ion channel activity
          voltage-gated calcium channel activity
          calcium channel activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050897 BC051630.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994284 NM_001130030.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994284 NM_177866.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994284 AK077145.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685147 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49063 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130718 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104560 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177959 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994284 NM_001130030.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994284 NM_177866.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994284 AK077145.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P234440 BC108974.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P234440 BC108975.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P234440 EU781510.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P234440 EU781511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P234440 NM_001130030.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23321667 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00994374 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104560 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177959 - -
          Whole Mouse Genome G4122A A_51_P234440 BC108974.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234440 BC108975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234440 EU781510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234440 EU781511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P234440 NM_001130030.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109906 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108952 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177959 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104560 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03884455 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03884439 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03884470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03884464 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03884460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23321553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03626732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03884460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03341665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03510856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04098679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03389418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03467002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03993688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03566601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03923106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23321657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03341666 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10517201 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517202 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517207 - -
          Affymetrix Mouse Gene 1.1 ST Array 10517199 - -
          Affymetrix Mouse Clariom S Array PSR0400030663.mm.1 TC0400003730.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030663.mm.1 NM_001130030.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030663.mm.1 NM_001130030.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030663.mm.1 NM_177866.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030663.mm.1 NM_177866.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030662.mm.1 TC0400003730.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030662.mm.1 NM_001130030.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030662.mm.1 NM_001130030.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030662.mm.1 NM_177866.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030662.mm.1 NM_177866.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 761216 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3440181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2591842 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2591842 - -
          mouse-6 v1.1 (Illumina) scl23695.12.1_184-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23695.12.1_184-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2991970 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2991970 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0