Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163269.1 | NP_001156741.1 | 1 - 2 | 55 | 60 | |
GenBank mRNA | AK131863.1 | - | 1 - 2 | 56 | 60 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |