Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01190972_m1
          See other 9330102E08RIK GE Assays ›
          Gene Symbol:
          9330102E08Rik
          Gene Name:
          RIKEN cDNA 9330102E08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 128169700 - 128183803 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01190972_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9330102E08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_077223.1 2 - 3 1232 102
          GenBank mRNA AK033854.1 - 2 - 3 1232 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tetraspanin 9 Tspan9 6720474K14Rik,
          9430079M16Rik,
          AU018597
          -26.1kb Sense Chr. 6 - 127961396 - 128143598 on NCBI Build GRCm38
          Downstream TEA domain family member 4 Tead4 ETFR-2,
          Etfr2,
          FR-19,
          Rtef1,
          TEAD-4,
          TEF-3,
          Tcf13r1,
          Tef3,
          Tefr,
          Tefr1,
          Tefr1a
          45.1kb Sense Chr. 6 - 128227143 - 128300813 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003192.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199717 AK033854.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199717 AW047626.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2199717 BB075161.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175998 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140359 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110072 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199717 AK033854.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199717 AW047626.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2199717 BB075161.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110072 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140359 - -
          Whole Mouse Genome G4122A A_52_P170173 AK033854.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139330 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140359 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05514820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05750743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05195265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05750752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05636748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05750747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05750739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05654309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05171467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05757558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05750737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05117649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24837608 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1439874_PM_at - -
          Mouse Genome 430 2.0 Array 1439874_at - -
          Mouse Genome 430 2.0 Array 1439874_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600024865.mm.1 TC0600003192.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024865.mm.1 NR_077223.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024866.mm.1 TC0600003192.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024866.mm.1 NR_077223.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024867.mm.1 TC0600003192.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024867.mm.1 NR_077223.1 Refseq
          Mouse Expression Set 430 1439874_at - -
          Mouse Expression Set 430 1439874_PM_at - -
          Murine Genome U74v2 Set 133789_at - -
          Murine Genome U74v2 Set 133789_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439874_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439874_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 487940 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103360301 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0