Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191050_m1
          See other GM19395 GE Assays ›
          Gene Symbol:
          Gm19395
          Gene Name:
          predicted gene, 19395
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 53280270 - 53283687 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191050_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm19395
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038015.1 2 - 3 407 125
          GenBank mRNA AK076832.1 - 2 - 3 397 125
          BC048600.1 - 2 - 3 393 125
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX633540.1 - 1 - 2 Chr. 10 - 53282607 - 53283687 [-] on Build GRCm38 125
          GenBankmRNA BU946220.1 - 2 - 3 396 Chr. 10 - 53280284 - 53283684 [+] on Build GRCm38 125
          GenBankmRNA BB615398.1 - 2 - 3 397 Chr. 10 - 53280281 - 53283683 [+] on Build GRCm38 125
          GenBankmRNA BF319476.1 - 1 - 2 Chr. 10 - 53282548 - 53283687 [-] on Build GRCm38 125
          GenBankmRNA AV264908.2 - 2 - 3 407 Chr. 10 - 53280287 - 53283686 [+] on Build GRCm38 125

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          36173
          Gm36173 -21.7kb Sense Chr. 10 - 53235189 - 53260752 on NCBI Build GRCm38
          Downstream phospholamban Pln Plb 54.0kb Sense Chr. 10 - 53337686 - 53345999 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107018 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107018 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107532 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08749325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08631271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27141473 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0