Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00004649.mm.1 |
TC0X00000703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004649.mm.1 |
NM_007927.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004657.mm.1 |
TC0X00000703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004657.mm.1 |
NM_007927.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004655.mm.1 |
TC0X00000703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004655.mm.1 |
NM_007927.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004656.mm.1 |
TC0X00000703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004656.mm.1 |
NM_007927.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004658.mm.1 |
TC0X00000703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004658.mm.1 |
NM_007927.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004651.mm.1 |
TC0X00000703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004651.mm.1 |
NM_007927.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004659.mm.1 |
TC0X00000703.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004659.mm.1 |
NM_007927.3 |
Refseq |