Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191882_m1
          See other LATS1 GE Assays ›
          Gene Symbol:
          Lats1
          Gene Name:
          large tumor suppressor
          Gene Aliases:
          AW208599
          Chromosome Location:
          Chr.10: 7681209 - 7716461 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191882_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lats1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010690.1 NP_034820.1 3 - 4 870 77
          GenBank mRNA AK038612.1 - 3 - 4 871 77
          BC158092.1 - 2 - 3 716 9088177 77
          BC158123.1 - 3 - 4 865 9088196 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000048.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          mitotic cell cycle
          sister chromatid segregation
          protein phosphorylation
          cell cycle
          hormone-mediated signaling pathway
          phosphorylation
          keratinocyte differentiation
          regulation of actin filament polymerization
          positive regulation of peptidyl-serine phosphorylation
          cellular protein localization
          hippo signaling
          regulation of protein-containing complex assembly
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          cytoplasmic sequestering of protein
          cell division
          negative regulation of canonical Wnt signaling pathway
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000000363.mm.1 TC1000000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000000363.mm.1 NM_010690.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000000363.mm.1 NM_010690.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000000361.mm.1 TC1000000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000000361.mm.1 NM_010690.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000000361.mm.1 NM_010690.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000000364.mm.1 TC1000000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000000364.mm.1 NM_010690.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000000364.mm.1 NM_010690.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000000365.mm.1 TC1000000048.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000000365.mm.1 NM_010690.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000000365.mm.1 NM_010690.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000000362.mm.1 TC1000000048.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0