Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400013241.mm.1 |
TC0400001662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013241.mm.1 |
NM_178257.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013241.mm.1 |
NM_178257.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013244.mm.1 |
TC0400001662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013244.mm.1 |
NM_178257.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013244.mm.1 |
NM_178257.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013240.mm.1 |
TC0400001662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013240.mm.1 |
NM_178257.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013240.mm.1 |
NM_178257.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013243.mm.1 |
TC0400001662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013243.mm.1 |
NM_178257.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013243.mm.1 |
NM_178257.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013242.mm.1 |
TC0400001662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013242.mm.1 |
NM_178257.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013242.mm.1 |
NM_178257.2 |
Refseq |