Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01197385_g1
          See other CRLF2 GE Assays ›
          Gene Symbol:
          Crlf2
          Gene Name:
          cytokine receptor-like factor 2
          Gene Aliases:
          CRLM2, Ly114, Tpte2, Tslpr
          Chromosome Location:
          Chr.5: 109554709 - 109559017 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01197385_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crlf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164735.1 NP_001158207.1 4 - 5 595 70
          NM_016715.4 NP_057924.3 4 - 5 595 70
          GenBank mRNA AB031333.1 - 4 - 5 543 70
          AB039945.1 - 4 - 5 491 70
          AF201963.1 - 4 - 5 646 70
          AF232936.1 - 4 - 5 559 70
          AK149928.1 - 4 - 5 596 70
          AK170466.1 - 1 - 2 457 70
          BC016580.1 - 4 - 5 673 2655857 70
          BC023788.1 - 4 - 5 583 5346748 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002969.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crlf2

          Gene Ontology Categories:

          Function(s) Process(es)

          immunoglobulin production involved in immunoglobulin-mediated immune response
          inflammatory response
          T-helper 2 cell cytokine production
          cytokine activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976360 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976362 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976361 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976348 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018847 AF232936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018847 AK010291.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018847 AK149928.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018847 AK170466.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018847 AB039945.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64087 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111769 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018847 AF232936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018847 AK010291.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018847 AK149928.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018847 AK170466.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018847 AB039945.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488340 BC023788.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488340 BC016580.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488340 AB031333.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P488340 AF201963.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23976360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23976366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23976363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23976362 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05691682 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111769 - -
          Whole Mouse Genome G4122A A_51_P488340 BC023788.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488340 BC016580.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488340 AB031333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P488340 AF201963.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136300 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111769 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04104973 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976360 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976361 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976362 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23976348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23976350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04702720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23976364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04702723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23976387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23976384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23976371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23976390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23976360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04457600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04261621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04702714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04702725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04702716 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532298 - -
          Affymetrix HT MG-430 PM Array Plate 1418097_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532289 - -
          Mouse Genome 430 2.0 Array 1418097_PM_a_at - -
          Mouse Genome 430 2.0 Array 1418097_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57914_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57914_at - -
          Affymetrix Mouse Clariom S Array PSR0500026280.mm.1 TC0500002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026280.mm.1 NM_001164735.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026280.mm.1 NM_001164735.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026280.mm.1 NM_016715.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026280.mm.1 NM_016715.4 Refseq
          Mouse Expression Set 430 1418097_a_at - -
          Mouse Expression Set 430 1418097_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1418097_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1418097_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1418097_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1418097_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2370358 - -
          Illumina mouse-6 v1.0 expression beadchip 2370358 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370358 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2514362 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2514362 - -
          mouse-6 v1.1 (Illumina) scl26240.7.1_74-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370358 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26240.7.1_74-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370358 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2514362 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0