Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01198326_m1
          See other 5930412G12RIK GE Assays ›
          Gene Symbol:
          5930412G12Rik
          Gene Name:
          RIKEN cDNA 5930412G12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 128579130 - 128600687 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01198326_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5930412G12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015517.2 5 - 6 2247 115
          GenBank mRNA AK031134.1 - 5 - 6 2248 115
          AK081926.1 - 5 - 6 2244 115
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB036132.2 - 3 - 4 544 Chr. 5 - 128579132 - 128585407 [-] on Build GRCm38 115
          GenBankmRNA BB377017.2 - 3 - 4 481 Chr. 5 - 128579104 - 128585348 [-] on Build GRCm38 115
          GenBankmRNA AI853239.1 - 1 - 2 Chr. 5 - 128579108 - 128582822 [+] on Build GRCm38 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane protein 132D Tmem132d C630028F04Rik -146.0kb Anti-sense Chr. 5 - 127781629 - 128433077 on NCBI Build GRCm38
          Downstream frizzled homolog 10 (Drosophila) Fzd10 Fz-10 18.3kb Sense Chr. 5 - 128601106 - 128604093 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003292.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114255 AK081926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114255 AK081926.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05740032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05740044 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05740069 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114255 AK081926.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05740044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24083902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24083908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24083910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24083918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04235662 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444013_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441338_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534014 - -
          Mouse Genome 430 2.0 Array 1444013_at - -
          Mouse Genome 430 2.0 Array 1441338_PM_at - -
          Mouse Genome 430 2.0 Array 1441338_at - -
          Mouse Genome 430 2.0 Array 1444013_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500029774.mm.1 TC0500003292.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500029774.mm.1 NR_015517.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500029773.mm.1 TC0500003292.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500029773.mm.1 NR_015517.2 Refseq
          Mouse Expression Set 430 1441338_at - -
          Mouse Expression Set 430 1444013_at - -
          Mouse Expression Set 430 1441338_PM_at - -
          Mouse Expression Set 430 1444013_PM_at - -
          Murine Genome U74v2 Set 136199_PM_at - -
          Murine Genome U74v2 Set 136199_at - -
          Affymetrix HT Mouse Genome 430B Array 1444013_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441338_at - -
          Affymetrix HT Mouse Genome 430B Array 1441338_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444013_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 511247 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 104610390 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222542 - -
          Illumina Sentrix mouse-6.v1 bead array 104610390 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104610390 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222542 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0