Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300009422.mm.1 |
TC0300001152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009422.mm.1 |
NM_001177770.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009422.mm.1 |
NM_001177770.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009422.mm.1 |
NM_001177771.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009422.mm.1 |
NM_001177771.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009422.mm.1 |
NM_145543.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009422.mm.1 |
NM_145543.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009423.mm.1 |
TC0300001152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009423.mm.1 |
NM_001177770.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009423.mm.1 |
NM_001177770.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009423.mm.1 |
NM_001177771.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009423.mm.1 |
NM_001177771.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009423.mm.1 |
NM_145543.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009423.mm.1 |
NM_145543.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009421.mm.1 |
TC0300001152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009421.mm.1 |
NM_001177770.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009421.mm.1 |
NM_001177770.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009421.mm.1 |
NM_001177771.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009421.mm.1 |
NM_001177771.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009421.mm.1 |
NM_145543.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009421.mm.1 |
NM_145543.2 |
Refseq |