Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01200460_g1
          See other LY6E GE Assays ›
          Gene Symbol:
          Ly6e
          Gene Name:
          lymphocyte antigen 6 complex, locus E
          Gene Aliases:
          9804, Ly67, RIG-E, Sca-2, TSA-1, Tsa1
          Chromosome Location:
          Chr.15: 74955051 - 74959905 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01200460_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly6e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164036.1 NP_001157508.1 2 - 3 752 80
          NM_001164037.1 NP_001157509.1 3 - 4 625 80
          NM_001164038.1 NP_001157510.1 3 - 4 541 80
          NM_001164039.1 NP_001157511.1 3 - 4 538 80
          NM_001164040.1 NP_001157512.1 2 - 3 335 80
          NM_008529.3 NP_032555.1 2 - 3 749 80
          GenBank mRNA AK014207.1 - 2 - 3 207 80
          BC002116.1 - 2 - 3 152 3490293 80
          BC003926.1 - 2 - 3 155 3485382 80
          BC005684.1 - 2 - 3 187 3155116 80
          BC019113.1 - 2 - 3 178 2651183 80
          BC037143.1 - 2 - 3 163 80
          BC040691.1 - 2 - 3 172 5025135 80
          BC053523.1 - 2 - 3 155 4205673 80
          BC080711.1 - 2 - 3 152 3969173 80
          CT010220.1 - 1 - 2 74 80
          CT010244.1 - 1 - 2 74 80
          U04268.1 - 1 - 2 117 80
          U09192.1 - 2 - 3 142 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000549.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly6e

          Gene Ontology Categories:

          Function(s)

          in utero embryonic development
          adrenal gland development
          organ growth
          norepinephrine metabolic process
          epinephrine secretion
          ventricular cardiac muscle tissue morphogenesis

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 TC1500000549.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164036.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164036.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164037.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164037.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164038.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164038.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164039.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164039.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164040.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_001164040.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_008529.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500003773.mm.1 NM_008529.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 927958 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0