Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025561.3 | NP_001020732.1 | 1 - 2 | 360 | 92 | |
NM_019700.4 | NP_062674.2 | 1 - 2 | 360 | 92 | ||
GenBank mRNA | AB041563.1 | - | 1 - 2 | 361 | 92 | |
AK089258.1 | - | 1 - 2 | 360 | 92 | ||
BC021446.1 | - | 1 - 2 | 236 | 5038201 | 92 | |
BC034359.1 | - | 1 - 2 | 331 | 3595162 | 92 | |
BU515425.1 | - | 1 - 2 | 332 | 92 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |