Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256115.1 | NP_001243044.1 | 5 - 6 | 568 | 101 | |
NM_001256116.1 | NP_001243045.1 | 4 - 5 | 505 | 101 | ||
NM_001256117.1 | NP_001243046.1 | 5 - 6 | 568 | 101 | ||
NM_001256118.1 | NP_001243047.1 | 4 - 5 | 505 | 101 | ||
NM_024451.2 | NP_077771.1 | 5 - 6 | 568 | 101 | ||
GenBank mRNA | AK014585.1 | - | 5 - 6 | 566 | 101 | |
AK036187.1 | - | 5 - 6 | 545 | 101 | ||
AK160281.1 | - | 5 - 6 | 526 | 101 | ||
AK161959.1 | - | 5 - 6 | 540 | 101 | ||
AK162533.1 | - | 5 - 6 | 539 | 101 | ||
AK167686.1 | - | 5 - 6 | 540 | 101 | ||
AK181636.1 | - | 2 - 3 | 92 | 101 | ||
AK194327.1 | - | 2 - 3 | 91 | 101 | ||
AK208681.1 | - | 3 - 4 | 219 | 101 | ||
AY682989.1 | - | 4 - 5 | 485 | 101 | ||
BC047928.1 | - | 5 - 6 | 535 | 5321879 | 101 | |
BC048156.1 | - | 5 - 6 | 537 | 5698402 | 101 | |
BQ551727.1 | - | 4 - 5 | 509 | 101 | ||
HQ402597.1 | - | 4 - 5 | 485 | 101 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |