Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500009618.mm.1 |
TC0500001130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009618.mm.1 |
NM_001281812.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009618.mm.1 |
NM_207279.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009618.mm.1 |
NM_207279.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009622.mm.1 |
TC0500001130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009622.mm.1 |
NM_001281812.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009622.mm.1 |
NM_207279.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009622.mm.1 |
NM_207279.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009620.mm.1 |
TC0500001130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009620.mm.1 |
NM_001281812.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009620.mm.1 |
NM_207279.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009620.mm.1 |
NM_207279.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009617.mm.1 |
TC0500001130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009617.mm.1 |
NM_001281812.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009617.mm.1 |
NM_207279.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009617.mm.1 |
NM_207279.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009621.mm.1 |
TC0500001130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009621.mm.1 |
NM_001281812.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009621.mm.1 |
NM_207279.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009621.mm.1 |
NM_207279.2 |
Refseq |