Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500032498.mm.1 |
TC0500003548.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032498.mm.1 |
NR_040424.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032498.mm.1 |
NR_040424.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032498.mm.1 |
NR_040425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032498.mm.1 |
NR_040425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032498.mm.1 |
NR_040426.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032498.mm.1 |
NR_040426.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032501.mm.1 |
TC0500003548.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032501.mm.1 |
NR_040424.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032501.mm.1 |
NR_040424.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032501.mm.1 |
NR_040425.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032501.mm.1 |
NR_040425.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032501.mm.1 |
NR_040426.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032501.mm.1 |
NR_040426.1 |
Refseq |