Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026718.2 | NP_080994.2 | 3 - 4 | 447 | 61 | |
GenBank mRNA | AK039432.1 | - | 3 - 4 | 640 | 61 | |
AK156539.1 | - | 3 - 4 | 640 | 61 | ||
BC027765.1 | - | 1 - 2 | 119 | 61 | ||
BC049187.1 | - | 3 - 4 | 626 | 61 | ||
BC061692.1 | - | 3 - 4 | 447 | 6308996 | 61 | |
BC108413.1 | - | 3 - 4 | 922 | 6487251 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |